Product Details

SNP ID
rs506688
Assay Type
Functionally tested
NCBI dbSNP Submissions
61
Location
Chr.1:155923959 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAGTTCATAAAATTACTATAAAA[C/T]GTATTAATATTGACATAGGTTTAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA0907 PubMed Links

Gene Details

Gene
KIAA0907
Gene Name
KIAA0907
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014949.3 Intron NP_055764.2
Gene
SCARNA4
Gene Name
small Cajal body-specific RNA 4
There are no transcripts associated with this gene.

Gene
SNORA80E
Gene Name
small nucleolar RNA, H/ACA box 80E
There are no transcripts associated with this gene.

View Full Product Details