Product Details
- SNP ID
-
rs9921169
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:3371261 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACCCTGTTGTTGATATGGACCCTAG[A/T]ATAGGATTGTGCCGTTATCCCTAGG
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MTRNR2L4
PubMed Links
Gene Details
- Gene
- MTRNR2L4
- Gene Name
- MT-RNR2-like 4
View Full Product Details