Product Details

SNP ID
rs12581486
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55246286 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACAGAAGAAAAGAGATGGCCATGG[C/T]AACAGGGGGAAACATGCAGAGCAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR6C74 PubMed Links

Gene Details

Gene
OR6C74
Gene Name
olfactory receptor family 6 subfamily C member 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005490.1 Intron NP_001005490.1
XM_017019126.1 Intron XP_016874615.1

View Full Product Details