Product Details
- SNP ID
-
rs11988053
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:66113265 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- ACACAGGCCGGCTCTGGGGCTCTGC[G/T]CTCCTCGGATTACGCATGCTCAGTG
- Phenotype
-
MIM: 606469
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TRIM55
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79140417] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TRIM55
- Gene Name
- tripartite motif containing 55
View Full Product Details