Product Details

SNP ID
rs1961957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122385275 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATTATCCTTTACACAGCTTATCT[A/G]GTGTTCCGGTAGAGAATGGACTGGA
Phenotype
MIM: 608017
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC58 PubMed Links

Gene Details

Gene
CCDC58
Gene Name
coiled-coil domain containing 58
There are no transcripts associated with this gene.

Gene
FAM162A
Gene Name
family with sequence similarity 162 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014367.3 Intron NP_055182.3

View Full Product Details