Product Details

SNP ID
rs1401078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:116711233 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTTCTAAGAAGTAAGGGCATGCA[G/T]ATTTATTTTCTCTGCAGCTTTAATG
Phenotype
MIM: 616057
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TUSC7 PubMed Links

Gene Details

Gene
TUSC7
Gene Name
tumor suppressor candidate 7 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details