Product Details
- SNP ID
-
rs1967376
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:70636027 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTAGATGGTATGAAACAAAACCAG[C/T]GCATCAGTAATCATGCAGAAAAAGA
- Phenotype
-
MIM: 606585
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ENAM
PubMed Links
Gene Details
- Gene
- ENAM
- Gene Name
- enamelin
View Full Product Details