Product Details

SNP ID
rs2496160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:13361695 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTATCTCAAACCCATTTGCTACA[G/T]TTTATATTTAAAGAAAATGAACTAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GFOD1 PubMed Links

Gene Details

Gene
GFOD1
Gene Name
glucose-fructose oxidoreductase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242628.1 4157 Intron NP_001229557.1
NM_001242629.1 4157 Intron NP_001229558.1
NM_001242630.1 4157 Intron NP_001229559.1
NM_018988.3 4157 Intron NP_061861.1
XM_011514699.2 4157 UTR 3 XP_011513001.1

View Full Product Details