Product Details

SNP ID
rs2048157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:48021329 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAAAGTCCCTCATTTTACAGATG[C/T]GGAAACTGAAACCAAGAGCCATTAC
Phenotype
MIM: 123825
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNGA1 PubMed Links

Gene Details

Gene
CNGA1
Gene Name
cyclic nucleotide gated channel alpha 1
There are no transcripts associated with this gene.

Gene
NIPAL1
Gene Name
NIPA like domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207330.2 Intron NP_997213.1
XM_017007784.1 Intron XP_016863273.1

View Full Product Details