Product Details

SNP ID
rs1886388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:11745281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTCCGCGGCGCACAATGGCCTC[A/G]GGGGCTGGAATCTTTTTATACAATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ECHDC3 PubMed Links

Gene Details

Gene
ECHDC3
Gene Name
enoyl-CoA hydratase domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024693.4 Intron NP_078969.2
XM_011519689.1 Intron XP_011517991.1

View Full Product Details