Product Details

SNP ID
rs2234710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30896514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCAAGGAATCCTGTCTCAAAAG[T/G]GGGGGTAGGATGAAATGTTTGGGGT
Phenotype
MIM: 605847 MIM: 600435
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
BCL7C PubMed Links

Gene Details

Gene
BCL7C
Gene Name
BCL tumor suppressor 7C
There are no transcripts associated with this gene.

Gene
CTF1
Gene Name
cardiotrophin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142544.1 Intron NP_001136016.1
NM_001330.3 Intron NP_001321.1
XM_011545759.2 Intron XP_011544061.1
XM_011545760.2 Intron XP_011544062.1
Gene
MIR762
Gene Name
microRNA 762
There are no transcripts associated with this gene.

Gene
MIR762HG
Gene Name
MIR762 host gene
There are no transcripts associated with this gene.

View Full Product Details