Product Details
- SNP ID
-
rs13381
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
45
- Location
-
Chr.1:232950710 on Build GRCh38
- Set Membership
-
JSNP
- Context Sequence [VIC/FAM]
- CGCAACCCCTACCCCCGCCCACCAG[C/T]ATGGCCCGGCACGTGTTCCTAACGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NTPCR
PubMed Links
Gene Details
- Gene
- NTPCR
- Gene Name
- nucleoside-triphosphatase, cancer-related
There are no transcripts associated with this gene.
View Full Product Details