Product Details

SNP ID
rs1132115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82321536 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCCGAGGGACTGAGGGATGCGCG[A/T]CCAGCCCCGGAGGGCGGCGTCCACC
Phenotype
MIM: 186820 MIM: 602602
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CD7 PubMed Links

Gene Details

Gene
CD7
Gene Name
CD7 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006137.6 1801 Intron NP_006128.1
XM_011523608.1 1801 Intron XP_011521910.1
XM_017025316.1 1801 Intron XP_016880805.1
Gene
SECTM1
Gene Name
secreted and transmembrane 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003004.2 1801 UTR 3 NP_002995.1
XM_005256392.3 1801 UTR 3 XP_005256449.1
XM_011523588.2 1801 UTR 3 XP_011521890.1

View Full Product Details