Product Details
- SNP ID
-
rs3391
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41690812 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGGCTAAGGACGATCAGCTGAAG[A/G]TTCATGGGTTTTAAGTGCTTGTGGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EIF1
PubMed Links
Gene Details
- Gene
- EIF1
- Gene Name
- eukaryotic translation initiation factor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005801.3 |
492 |
Missense Mutation |
ATT,GTT |
I110V |
NP_005792.1 |
View Full Product Details