Product Details
- SNP ID
-
rs11882
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:22900960 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGCCTCCCTCCATCCTCCTACCT[C/T]GGGCCAAAGGGAGGAAATGGGAGGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RBM23
PubMed Links
Gene Details
- Gene
- RBM23
- Gene Name
- RNA binding motif protein 23
View Full Product Details