Product Details

SNP ID
rs14681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8879094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGACATTAAGACAGGGGAGAGAC[G/T]GCCAGGCAAGGTAGAAGCTGGCTCT
Phenotype
MIM: 611317 MIM: 611462
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PIK3R5 PubMed Links

Gene Details

Gene
PIK3R5
Gene Name
phosphoinositide-3-kinase regulatory subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142633.2 4143 UTR 3 NP_001136105.1
NM_001251851.1 4143 UTR 3 NP_001238780.1
NM_001251852.1 4143 UTR 3 NP_001238781.1
NM_001251853.1 4143 UTR 3 NP_001238782.1
NM_001251855.1 4143 UTR 3 NP_001238784.1
NM_014308.3 4143 UTR 3 NP_055123.2
XM_005256580.3 4143 UTR 3 XP_005256637.1
Gene
PIK3R6
Gene Name
phosphoinositide-3-kinase regulatory subunit 6
There are no transcripts associated with this gene.

View Full Product Details