Product Details

SNP ID
rs13340
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:145992739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATCTCCTAACACAGGGCAGGAA[G/A]GGGGTTCAAAGGGCTTTTATGAAGA
Phenotype
MIM: 606599
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
TXNIP PubMed Links

Gene Details

Gene
TXNIP
Gene Name
thioredoxin interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001313972.1 2731 UTR 3 NP_001300901.1
NM_006472.5 2731 UTR 3 NP_006463.3
XM_017000085.1 2731 UTR 3 XP_016855574.1

View Full Product Details