Product Details

SNP ID
rs1984358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:64081849 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAAGCTGCATCCTCCAGGTGGAC[A/G]TGACCACATGGTTTGGCAAGCCTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00305 PubMed Links

Gene Details

Gene
LINC00305
Gene Name
long intergenic non-protein coding RNA 305
There are no transcripts associated with this gene.

View Full Product Details