Product Details

SNP ID
rs10927787
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:15457721 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTCCCCTCCCCACTGCCCATAGG[C/T]AGCAAATATAGTATTACTCTATTAA
Phenotype
MIM: 609443 MIM: 601405
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CELA2A PubMed Links

Gene Details

Gene
CELA2A
Gene Name
chymotrypsin like elastase family member 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033440.2 Intron NP_254275.1
Gene
CTRC
Gene Name
chymotrypsin C
There are no transcripts associated with this gene.

View Full Product Details