Product Details

SNP ID
rs2234920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:32801104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCTGGGAGAGCACAGAAAACAG[A/T]CTTCTATCATATATCATAGCCAGCT
Phenotype
MIM: 602705 MIM: 188826
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SYN3 PubMed Links

Gene Details

Gene
SYN3
Gene Name
synapsin III
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135774.1 289 Intron NP_001129246.1
NM_003490.3 289 Intron NP_003481.3
NM_133633.2 289 Intron NP_598344.2
XM_011530405.2 289 Intron XP_011528707.1
XM_011530406.2 289 Intron XP_011528708.1
XM_011530407.2 289 Intron XP_011528709.1
XM_011530408.1 289 Intron XP_011528710.1
XM_011530410.2 289 Intron XP_011528712.1
XM_017028960.1 289 Intron XP_016884449.1
XM_017028961.1 289 Intron XP_016884450.1
XM_017028962.1 289 Intron XP_016884451.1
XM_017028963.1 289 Intron XP_016884452.1
XM_017028964.1 289 Intron XP_016884453.1
XM_017028965.1 289 Intron XP_016884454.1
Gene
TIMP3
Gene Name
TIMP metallopeptidase inhibitor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000362.4 289 UTR 5 NP_000353.1

View Full Product Details