Product Details

SNP ID
rs3892537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:129738628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTGAGCTTCCTGTGGGCCCCTC[A/C]CACCCACACCTGAGCCAGAGAGAAG
Phenotype
MIM: 605172
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PTGES PubMed Links

Gene Details

Gene
PTGES
Gene Name
prostaglandin E synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004878.4 1856 UTR 3 NP_004869.1
XM_011519210.2 1856 UTR 3 XP_011517512.1

View Full Product Details