Product Details

SNP ID
rs2026196
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:112489584 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTCCAGAGAACCTTACATATTTTCA[A/G]CTGATTTCCTTTCTGGTAAGTTGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf147 PubMed Links

Gene Details

Gene
C9orf147
Gene Name
chromosome 9 open reading frame 147
There are no transcripts associated with this gene.

Gene
KIAA1958
Gene Name
KIAA1958
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287036.1 Intron NP_001273965.1
NM_001287038.1 Intron NP_001273967.1
NM_133465.3 Intron NP_597722.1
XM_011518311.2 Intron XP_011516613.1

View Full Product Details