Product Details
- SNP ID
-
rs1890101
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:128559474 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGTGACTCCTGTGATAGGAGGCA[G/T]AACAGCACCATACACTCGAATCCTG
- Phenotype
-
MIM: 182810
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SPTAN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75428787] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SPTAN1
- Gene Name
- spectrin alpha, non-erythrocytic 1
View Full Product Details