Product Details

SNP ID
rs6053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:154563022 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGATCTCTCAGTTAAGTCTACATG[A/G]AAAGGATGGTTTCTTGGAGCTTCCA
Phenotype
MIM: 134830
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGB PubMed Links

Gene Details

Gene
FGB
Gene Name
fibrinogen beta chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184741.1 43 Missense Mutation AAA,GAA K2E NP_001171670.1
NM_005141.4 43 Missense Mutation AAA,GAA K2E NP_005132.2

View Full Product Details