Product Details
- SNP ID
-
rs2066864
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:154604543 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTTGATATAATGAAAATAATTTAC[A/G]AAGACAAAATAAATGACAAGTGGTC
- Phenotype
-
MIM: 134850
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FGG
PubMed Links
Gene Details
- Gene
- FGG
- Gene Name
- fibrinogen gamma chain
View Full Product Details