Product Details

SNP ID
rs1790419
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:50238007 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTATTAGGATTTAAGGTTGGTG[C/T]CAATTTTTTTAAGTGTTTCAAATTT
Phenotype
MIM: 614759
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFAP53 PubMed Links

Gene Details

Gene
CFAP53
Gene Name
cilia and flagella associated protein 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145020.4 Intron NP_659457.2

View Full Product Details