Product Details
- SNP ID
-
rs12465520
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:219264699 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGTTCTAGACATGATGGAGTAGGA[C/T]GGCACTAGATTTCATCACACTACTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TUBA4B
PubMed Links
Gene Details
- Gene
- TUBA4B
- Gene Name
- tubulin alpha 4b
There are no transcripts associated with this gene.
View Full Product Details