Product Details

SNP ID
rs9612385
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23589994 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCCTGATAGTTTGGCTCGATCCCC[A/G]GATGTCTATTGTTCTCTGGCATAAG
Phenotype
MIM: 146770
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DRICH1 PubMed Links

Gene Details

Gene
DRICH1
Gene Name
aspartate rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016449.3 Intron NP_057533.2
XM_011530206.1 Intron XP_011528508.1
XM_011530207.2 Intron XP_011528509.1
XM_011530208.2 Intron XP_011528510.1
XM_011530209.2 Intron XP_011528511.1
XM_011530210.2 Intron XP_011528512.1
XM_017028813.1 Intron XP_016884302.1
Gene
IGLL1
Gene Name
immunoglobulin lambda like polypeptide 1
There are no transcripts associated with this gene.

View Full Product Details