Product Details

SNP ID
rs2272295
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:47475414 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTCTGGGAAGAGTGGGTGCATGGGT[A/G]CTTAAGAGGCTGCATTCTCAGCAGG
Phenotype
MIM: 615620 MIM: 601901
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KPTN PubMed Links

Gene Details

Gene
KPTN
Gene Name
kaptin, actin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291296.1 1249 UTR 3 NP_001278225.1
NM_007059.3 1249 UTR 3 NP_008990.2
XM_011526399.2 1249 UTR 3 XP_011524701.1
XM_017026226.1 1249 UTR 3 XP_016881715.1
XM_017026227.1 1249 UTR 3 XP_016881716.1
XM_017026228.1 1249 UTR 3 XP_016881717.1
XM_017026229.1 1249 Intron XP_016881718.1
Gene
NAPA-AS1
Gene Name
NAPA antisense RNA 1
There are no transcripts associated with this gene.

Gene
SLC8A2
Gene Name
solute carrier family 8 member A2
There are no transcripts associated with this gene.

View Full Product Details