Product Details
- SNP ID
-
rs2292832
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:240456086 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GGGACGGGGGCTGTGCTGGGGCAGC[C/T]GGAACAACGCAGGTCGCCGGGCCGG
- Phenotype
-
MIM: 600395
MIM: 615209
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GPC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs71428439] are located under a probe and SNP(s) [rs71428440] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GPC1
- Gene Name
- glypican 1
- Gene
- MIR149
- Gene Name
- microRNA 149
There are no transcripts associated with this gene.
- Gene
- PP14571
- Gene Name
- uncharacterized LOC100130449
There are no transcripts associated with this gene.
View Full Product Details