Product Details

SNP ID
rs12618600
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:73663555 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TATTCAGATCTAAGAGATAATTTTC[C/T]AGACATTTTTCTGATTTCTTTCTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALMS1P1 PubMed Links

Gene Details

Gene
ALMS1P1
Gene Name
ALMS1, centrosome and basal body associated protein pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details