Product Details

SNP ID
rs11126411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73648575 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAAGAGCAGTCTGACCTCCACAT[A/G]GAGCCCCCTCATTTCCACTCCTCAC
Phenotype
MIM: 606716
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALMS1P1 PubMed Links

Gene Details

Gene
ALMS1P1
Gene Name
ALMS1, centrosome and basal body associated protein pseudogene 1
There are no transcripts associated with this gene.

Gene
NAT8
Gene Name
N-acetyltransferase 8 (putative)
There are no transcripts associated with this gene.

View Full Product Details