Product Details

SNP ID
rs2811956
Assay Type
Functionally tested
NCBI dbSNP Submissions
53
Location
Chr.1:23630828 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTGAGCTGGTGGGAGGTGTAGGGG[A/G]GTTCACTGTGTTATTCTTTCAACCT
Phenotype
MIM: 607305
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MDS2 PubMed Links

Gene Details

Gene
MDS2
Gene Name
myelodysplastic syndrome 2 translocation associated
There are no transcripts associated with this gene.

View Full Product Details