Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_152778.2 | 2750 | UTR 3 | NP_689991.1 | ||
XM_005262893.1 | 2750 | UTR 3 | XP_005262950.1 | ||
XM_005262896.1 | 2750 | UTR 3 | XP_005262953.1 | ||
XM_005262897.2 | 2750 | UTR 3 | XP_005262954.1 | ||
XM_005262898.2 | 2750 | UTR 3 | XP_005262955.1 | ||
XM_011531830.1 | 2750 | UTR 3 | XP_011530132.1 | ||
XM_011531831.1 | 2750 | UTR 3 | XP_011530133.1 | ||
XM_011531832.1 | 2750 | UTR 3 | XP_011530134.1 | ||
XM_017007989.1 | 2750 | UTR 3 | XP_016863478.1 |