Product Details

SNP ID
rs35885016
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:139309919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCCTGGGTCATTATTGGGGCTCA[A/G]ACTAGTTATTTGATGATAATAGAAT
Phenotype
MIM: 608000 MIM: 603844
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAA15 PubMed Links

Gene Details

Gene
NAA15
Gene Name
N(alpha)-acetyltransferase 15, NatA auxiliary subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_057175.3 Intron NP_476516.1
XM_005263236.2 Intron XP_005263293.1
Gene
NDUFC1
Gene Name
NADH:ubiquinone oxidoreductase subunit C1
There are no transcripts associated with this gene.

View Full Product Details