Product Details

SNP ID
rs5030944
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:69225907 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGTGACGTACGTGGACGCACTCT[A/G]TAGACAGTAGCACGTGTTCTTCACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HKDC1 PubMed Links

Gene Details

Gene
HKDC1
Gene Name
hexokinase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025130.3 Intron NP_079406.3
XM_011540195.2 Intron XP_011538497.1
Gene
LOC101928994
Gene Name
uncharacterized LOC101928994
There are no transcripts associated with this gene.

View Full Product Details