Product Details

SNP ID
rs2024241
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:74244546 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGTTGAGGCTGCATTTGACCTTCCG[A/G]TGAATCTTCTAACTTTGAAAGGCAT
Phenotype
MIM: 142993
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VSX2 PubMed Links

Gene Details

Gene
VSX2
Gene Name
visual system homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182894.2 Intron NP_878314.1

View Full Product Details