Product Details

SNP ID
rs1133647
Assay Type
Functionally tested
NCBI dbSNP Submissions
50
Location
Chr.1:1081961 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
CCCGAAGGCCAGGCCACCCCTCAGA[T/G]GGGGGTCCCATGCTAAAGCAGACGG
Phenotype
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
C1orf159 PubMed Links

Gene Details

Gene
C1orf159
Gene Name
chromosome 1 open reading frame 159
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017891.4 2365 UTR 3 NP_060361.4
XM_005244765.2 2365 UTR 3 XP_005244822.1
XM_006710717.2 2365 UTR 3 XP_006710780.1
XM_006710718.2 2365 UTR 3 XP_006710781.1
XM_011541651.2 2365 UTR 3 XP_011539953.1
XM_011541652.2 2365 UTR 3 XP_011539954.1
XM_011541653.2 2365 UTR 3 XP_011539955.1
XM_011541655.2 2365 UTR 3 XP_011539957.1
XM_011541656.2 2365 UTR 3 XP_011539958.1
XM_011541658.2 2365 UTR 3 XP_011539960.1
XM_011541659.2 2365 UTR 3 XP_011539961.1
XM_011541660.2 2365 UTR 3 XP_011539962.1
XM_011541661.2 2365 UTR 3 XP_011539963.1
XM_011541662.2 2365 UTR 3 XP_011539964.1
XM_011541663.2 2365 UTR 3 XP_011539965.1
XM_011541665.2 2365 UTR 3 XP_011539967.1
XM_017001572.1 2365 UTR 3 XP_016857061.1
XM_017001573.1 2365 UTR 3 XP_016857062.1
XM_017001574.1 2365 UTR 3 XP_016857063.1
XM_017001575.1 2365 UTR 3 XP_016857064.1
XM_017001576.1 2365 UTR 3 XP_016857065.1
XM_017001577.1 2365 UTR 3 XP_016857066.1
XM_017001578.1 2365 UTR 3 XP_016857067.1
XM_017001579.1 2365 UTR 3 XP_016857068.1
XM_017001580.1 2365 UTR 3 XP_016857069.1
Gene
RNF223
Gene Name
ring finger protein 223
There are no transcripts associated with this gene.

View Full Product Details