Product Details

SNP ID
rs28383572
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:159588360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCAAAGTTATCGAAAAGTTCCC[A/G]GCTCCAGTGCACATCTGTGTGAGCT
Phenotype
MIM: 104770
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APCS PubMed Links

Gene Details

Gene
APCS
Gene Name
amyloid P component, serum
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001639.3 420 Silent Mutation CCA,CCG P108P NP_001630.1

View Full Product Details