Product Details

SNP ID
rs17179362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29403970 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATGGAGTAATTAAAAGATCATTT[C/T]ATCTTCAGCTTAGGTAGTGAAAAAG
Phenotype
MIM: 610266
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAOK1 PubMed Links

Gene Details

Gene
TAOK1
Gene Name
TAO kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020791.2 Intron NP_065842.1
NM_025142.1 Intron NP_079418.1
XM_011525060.2 Intron XP_011523362.1

View Full Product Details