Product Details

SNP ID
rs3922357
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32992015 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTCCTGGGGCTGTTAACAGGGGC[C/G]CTGGGTTGGTCTGGCCAGAGACCAG
Phenotype
MIM: 612749
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SPACA3 PubMed Links

Gene Details

Gene
SPACA3
Gene Name
sperm acrosome associated 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317225.1 Intron NP_001304154.1
NM_001317226.1 Intron NP_001304155.1
NM_173847.4 Intron NP_776246.1

View Full Product Details