Product Details

SNP ID
rs8106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:175178369 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGACAAGGCAAATCCCAGGATAGC[A/G]TATGAGAACAGCTGCTGCTTCAGCG
Phenotype
MIM: 602736
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP5G3 PubMed Links

Gene Details

Gene
ATP5G3
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C3 (subunit 9)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002258.4 1182 Silent Mutation TAC,TAT Y116Y NP_001002258.1
NM_001190329.1 1182 UTR 3 NP_001177258.1
NM_001689.4 1182 Silent Mutation TAC,TAT Y116Y NP_001680.1

View Full Product Details