Product Details

SNP ID
rs12410777
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:201893288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATATGACCTTTTCTCCCCCTCCT[C/T]CCACAAATATTGCCTAGAAGAGCCC
Phenotype
MIM: 602715
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IPO9 PubMed Links

Gene Details

Gene
IPO9
Gene Name
importin 9
There are no transcripts associated with this gene.

Gene
LMOD1
Gene Name
leiomodin 1
There are no transcripts associated with this gene.

Gene
SHISA4
Gene Name
shisa family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198149.2 Intron NP_937792.2
XM_017000419.1 Intron XP_016855908.1

View Full Product Details