Product Details

SNP ID
rs1801575
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:11106643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTGCATTTAGTTGAGTATTTGT[C/T]CTGCTCATAATTTCCAATATGTACC
Phenotype
MIM: 605960 MIM: 601231
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOSC10 PubMed Links

Gene Details

Gene
EXOSC10
Gene Name
exosome component 10
There are no transcripts associated with this gene.

Gene
LOC105376736
Gene Name
uncharacterized LOC105376736
There are no transcripts associated with this gene.

Gene
MTOR
Gene Name
mechanistic target of rapamycin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004958.3 8076 UTR 3 NP_004949.1
XM_005263438.2 8076 UTR 3 XP_005263495.1
XM_011541166.2 8076 Intron XP_011539468.1
XM_017000900.1 8076 UTR 3 XP_016856389.1
XM_017000901.1 8076 UTR 3 XP_016856390.1
XM_017000902.1 8076 Intron XP_016856391.1

View Full Product Details