Product Details
- SNP ID
-
rs7099737
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:15053123 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATATGATAGGCTCTTTTAATATGTC[A/G]TTTTCTTTCTTATTTTATTTCAGGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OLAH
PubMed Links
Gene Details
- Gene
- OLAH
- Gene Name
- oleoyl-ACP hydrolase
View Full Product Details