Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001164443.1 | Intron | NP_001157915.1 | |||
XM_011543298.2 | Intron | XP_011541600.1 | |||
XM_011543299.1 | Intron | XP_011541601.1 | |||
XM_011543300.2 | Intron | XP_011541602.1 | |||
XM_011543301.2 | Intron | XP_011541603.1 | |||
XM_011543302.2 | Intron | XP_011541604.1 | |||
XM_017009317.1 | Intron | XP_016864806.1 | |||
XM_017009318.1 | Intron | XP_016864807.1 | |||
XM_017009319.1 | Intron | XP_016864808.1 | |||
XM_017009320.1 | Intron | XP_016864809.1 | |||
XM_017009321.1 | Intron | XP_016864810.1 | |||
XM_017009322.1 | Intron | XP_016864811.1 |