Product Details

SNP ID
rs2021621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:31524767 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGCTATATTATTCTTCTACATCAT[A/G]CAATAAATTCTGAGCTAACCAGCCT
Phenotype
MIM: 606985
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ELP4 PubMed Links

Gene Details

Gene
ELP4
Gene Name
elongator acetyltransferase complex subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288725.1 Intron NP_001275654.1
NM_001288726.1 Intron NP_001275655.1
NM_019040.4 Intron NP_061913.3

View Full Product Details