Product Details
- SNP ID
-
rs10736996
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:5049067 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAAAGAGAAAAAGTCAACCTTTGTT[G/T]TGCATAGAAGTCTGGTTGCATGGTC
- Phenotype
-
MIM: 603966
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AKR1C3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79629670] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AKR1C3
- Gene Name
- aldo-keto reductase family 1, member C3
View Full Product Details