Product Details

SNP ID
rs1883784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:38606495 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTCTTGACCAGACATTATTGTTC[A/G]TTTTTAGTCTTGGCTAATTGGATTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP40 PubMed Links

Gene Details

Gene
ARHGAP40
Gene Name
Rho GTPase activating protein 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164431.1 Intron NP_001157903.1

View Full Product Details