Product Details

SNP ID
rs7952435
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:64279400 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCCTAACTGGACTGAGACAAGCACA[T/A]CTCTGGTTCAGACTCCAAGTCCCTG
Phenotype
MIM: 603167 MIM: 600230
Polymorphism
T/A, Transversion Substitution
Allele Nomenclature
Literature Links
BAD PubMed Links

Gene Details

Gene
BAD
Gene Name
BCL2 associated agonist of cell death
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004322.3 Intron NP_004313.1
NM_032989.2 Intron NP_116784.1
Gene
GPR137
Gene Name
G protein-coupled receptor 137
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170726.1 Intron NP_001164197.1
NM_001170880.1 Intron NP_001164351.1
NM_001170881.1 Intron NP_001164352.1
NM_001177358.1 Intron NP_001170829.1
NM_020155.3 Intron NP_064540.3
XM_005274100.2 Intron XP_005274157.1
XM_005274101.2 Intron XP_005274158.1
XM_005274102.2 Intron XP_005274159.1
XM_005274104.2 Intron XP_005274161.1
XM_011545168.2 Intron XP_011543470.1
XM_011545169.1 Intron XP_011543471.1
XM_011545170.2 Intron XP_011543472.1
XM_011545171.2 Intron XP_011543473.1
XM_011545172.2 Intron XP_011543474.1
XM_017018014.1 Intron XP_016873503.1
XM_017018015.1 Intron XP_016873504.1
XM_017018016.1 Intron XP_016873505.1
Gene
PLCB3
Gene Name
phospholipase C beta 3
There are no transcripts associated with this gene.

View Full Product Details